Please login to your online account to display your discounted pricing

Eppendorf™ LoBind PCR Plates

Manufacturer: eppendorf™  925000101

 View all options for this product

Catalog No. E925000101


Description & Specifications


Color White
For Use With (Application) Protein, peptide, antibody or virus samples
Material Polypropylene
No. of Wells 384
Surface Coating Siliconization
Type 384 Deep-Well Microplate
Volume (Metric) Well 200μL