Please login to your online account to display your discounted pricing

Eppendorf™ LoBind PCR Plates


Manufacturer: eppendorf™  925000096

 View more versions of this product

Catalog No. E925000096

  • / Pack
 

Description & Specifications

Specifications

Color White
For Use With (Application) Protein, peptide, antibody or virus samples
Material Polypropylene
No. of Wells 96
Surface Coating Siliconization
Type 96 Deep-Well Microplate
Volume (Metric) Well 500μL