Please login to your online account to display your discounted pricing

Eppendorf™ LoBind PCR Plates


Manufacturer: eppendorf™  925000095

 View more versions of this product

Catalog No. E925000095

  • / Pack
 

Description & Specifications

Specifications

No. of Wells 96
Color White
Type 96 Deep-Well Microplate
Material Polypropylene
Surface Coating Siliconization
For Use With (Application) Protein, peptide, antibody or virus samples
Volume (Metric) Well 2000μL