Eppendorf™ IsoTherm Systems: Starter Sets
Protect temperature-sensitive biochemical and microbiological samples while working at the lab bench
Manufacturer: Eppendorf™ 022510002
Includes: Starter Sets (Catalog Nos. 0540510 and 0540511) include:IsoSafe Transport Box, 0°C IsoPack, -21°C IsoPack, IsoRack for either 0.5mL tubes or 2.0 and 1.5mL tubes.
Also available with just the IsoPack (Catalog Nos. 0540516, 0540517, 0540519, 0540520) and IsoRack (Catalog Nos. 0540513, 0540519, 0540520, 0540522) for samples that Don't require the extended cooling time of the complete system, as well as the Isosafe (Catalog No. 0540518) for isotherm systems.
Description
For use with Eppendorf IsoTherm Systems
Specifications
Isotherm Starters Set | |
IsoSafe, IsoRack, 2 IsoPacks | |
24.0 |
24 x 0.5mL microtube, white isopack maintains 0°C for up to 6 Hours, blue isopack maintains -21°C for up to 3 Hours | |
Ideal for cooling, transporting and storing deep-frozen samples |