Eppendorf™ epTIPS in Racks
Maximum biological purity to satisfy the most stringent demands in medicine, the pharmaceutical and food industries, molecular biology and cell technology
Manufacturer:  Eppendorf™ 022491989
 View more versions of this product
Catalog No. 05-403-71
Description
Description
  • Continuous control of each batch by an independent laboratory
  • Packed in racks of 96, 48 or 24 tips, depending upon tip volume
Specifications
Specifications
Rack
100 to 5000μL
Violet
Plastic
12cm
Reliable, reproducible, high-precision pipetting for standard laboratory use
Extended Length,Finely Tapered,Universal Fit
Non-sterile
Yes
4.7 in.
ISO 9001
Documents