Eppendorf™ epTIPS in Racks
Maximum biological purity to satisfy the most stringent demands in medicine, the pharmaceutical and food industries, molecular biology and cell technology
Manufacturer:  Eppendorf™ 022491938
 View more versions of this product
Catalog No. 05-403-66
Description
Description
  • Continuous control of each batch by an independent laboratory
  • Packed in racks of 96, 48 or 24 tips, depending upon tip volume
Specifications
Specifications
Rack
2 to 200μL
Yellow
Plastic
5.3cm
Reliable, reproducible, high-precision pipetting for standard laboratory use
Extended Length, Finely Tapered, Universal Fit
Non-sterile
Yes
2.08 in.
ISO 9001
Documents