Eppendorf™ epTIPS™ PCR Clean
For all molecular biology applications
Manufacturer:  Eppendorf™ 022491725
 View more versions of this product
Catalog No. 05-403-48
Description
Description
Features
  • Free of human DNA-, DNase-, RNase- and PCR inhibitors
  • Drop contour design at tip provides better tip sealing
  • Finely tapered and extended; ideal for small-volume pipetting Fine graduations make it easy to check volume Soft, flexible tip cone provides better seal to pipetter Manufactured under strict quality standards in ISO-9001-certified production process
Packaging
  • Refill trays offer simple, contamination-free handling
  • Trays are color coded for easy volume identification
  • Environmentally friendly—all components of refills completely recyclable
  • Refill trays can be autoclaved intact and stored for future use
Specifications
Specifications
Refill
0.5 to 20μL
Gray
1.81 in.
ISO 9001
Universal Fit
Sterile
Yes
4.6cm
Documents