Eppendorf™ epTIPS™ LoRetention Tips
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Manufacturer:  Eppendorf™ 0030072251
 View more versions of this product
Catalog No. 13-683-501
Description
Description
  • Homogenous surface achieved through treatment at the molecular level — Pearl Effect Technology
  • Tips are uncoated and free from additives
  • Ultra-homogenous surface for maximum reproducibility
  • Ultra-hydrophobic surface for minimum loss of sample
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable, high chemical resistance
  • Five-year shelf life
Specifications
Specifications
Low Retention
Dark Gray
1.3 in.
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
0.1 to 10μL
Yes
3.4cm
Documents