Please login to your online account to display your discounted pricing
Eppendorf™ epTIPS™ Filter Tips
Ideal for small volume pipetting
Manufacturer: eppendorf™  022491229
 View more versions of this product
Catalog No. 05-403-16
  • / Case of 960 Each
Description & Specifications
Specifications
Product Type Drop Contour Tip
Volume (Metric) 0.5 to 20μL
Sterility Sterile
Color Gray
Material Polyethylene
Packaging Rack
Length (English) 1.81 in.
Length (Metric) 4.6cm
Certifications/Compliance ISO 9001