Eppendorf™ epTIPS™ Filter Tips
Ideal for small volume pipetting
Manufacturer:  Eppendorf™ 022491211
 View more versions of this product
Catalog No. 05-403-15
Description
Description
  • Drop contour design with filter
  • Finely tapered and extended
  • Fine graduations make it easy to check volume
  • Soft, flexible tip cone provides better seal to pipetter
  • Integrated filter of hydrophobic polyethylene
  • No self-sealing additives
  • Sample can be easily recovered if wet
Order Info
Packaged in racks of 96 tips.
Certifications
Batch-related certificates of purity are available upon request.
Specifications
Specifications
Rack
Sterile
PE (Polyethylene)
4cm
0.1 to 10μL
Gray
1.57 in.
ISO 9001
Documents