Eppendorf™ epTIPS™ Filter Tips
Ideal for small volume pipetting
$161.30 - $348.00
Specifications
Certifications/Compliance ISO 9001
Material PE (Polyethylene)
Format Rack
View More Specs
Products 3
Catalog Number Mfr. No. Sterility Volume (Metric) Length (English) Color Price Quantity  
Catalog Number Mfr. No. Sterility Volume (Metric) Length (English) Color Price Quantity  
05403120
View Documents
Eppendorf™
022491288
Sterile 1 to 10 mL 9.56 in. Turquoise Pack of 100 Each for $161.30
05-403-14
View Documents
Eppendorf™
022491202
Sterile 0.1 to 10 μL 1.3 in. Gray Case of 960 Each for $348.00
05-403-18
View Documents
Eppendorf™
022491237
Sterile 2 to 100 μL 2.16 in. Yellow Case of 960 Each for $325.59
Description
Description
  • Drop contour design with filter
  • Finely tapered and extended
  • Fine graduations make it easy to check volume
  • Soft, flexible tip cone provides better seal to pipetter
  • Integrated filter of hydrophobic polyethylene
  • No self-sealing additives
  • Sample can be easily recovered if wet
Order Info
Packaged in racks of 96 tips.
Certifications
Batch-related certificates of purity are available upon request.
Specifications
Specifications
ISO 9001
PE (Polyethylene)
Rack