missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ epTIPS™ Boxes and Refill Trays

Contamination-free transfer of trays to the working box

Manufacturer:  Eppendorf™ 022491351

 View more versions of this product

Catalog No. 05-403-26



Description

Description

  • System optimized for use with multi-channel pipettes
  • Refill trays can be autoclaved in advance
  • Tips can be used straight from the refill packaging
  • Reduced waste
  • All components 100% recyclable
  • Space-saving packaging
  • Color-coded trays for simple identification of tips and matching pipette

Specifications

Specifications

Extended Length, Finely Tapered, Universal Fit
Sterile
Yes
2.79 in.
ISO 9001
50 to 1000μL
Blue
Polypropylene
7.1cm
Reliable, reproducible, high-precision pipetting for standard laboratory use
Documents