Eppendorf™ epTIPS™ in Bags
The original, high-quality eppendorf pipette tips
Manufacturer:  Eppendorf™ 022492047
 View more versions of this product
Catalog No. 05-403-58
Description
Description
The original, high-quality eppendorf pipette tips
  • Available in all tip sizes from 10μl to 10mL
  • 200μl, 300μl and 1,000μl tips are color coded yellow and blue
  • Packed in re-sealable bags
Specifications
Specifications
Extended Length,Finely Tapered,Universal Fit
Non-Sterile
Yes
2.08 in.
ISO 9001
20 to 300μL
Orange
Plastic
5.3cm
Reliable, reproducible, high-precision pipetting for standard laboratory use
Documents