Please login to your online account to display your discounted pricing
Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax Filter Pipet Tips
Tips with liquid blocking technology provide additional protection against over pipetting
$232.78 - $232.78
Specifications
Material Polyethylene
Product Type Filter Pipet Tips
Sterility Sterile
View More Specs
Products
Catalog Number Mfr. No. Length (Metric) Color Volume (Metric) Price Quantity    
13-683-753
eppendorf™
0030077830
5.5cm Yellow 2 to 200μL Case of 960 Each for $232.78
Description & Specifications
Specifications
Material Polyethylene
Product Type Filter Pipet Tips
Sterility Sterile
  • Sterile tips made of polyethylene
  • White and violet filter layers in the tip retain aerosols and biomolecules
  • Purple filter layer blocks liquids, binds aerosols and does not allow any liquid to pass through
  • Hydrophobic white layer repels drops, splashes and binds aerosols
  • Free of PCR inhibiting additives
  • Provide protection against contamination of pipet and sample
  • Color coded tip boxes simplify matching pipetes with their tips
PCR and microbiology procedures
10 Racks x 96 tips
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.