Eppendorf™ Eppendorf™ Electronic Pipet Charging stands
Store Eppendorf pipettes and/or liquid handling instruments in a comfortable and safe way with this pipette carousel. Eppendorf™ Electronic Pipet Charging Stands charge electronic pipettes in an uncomplicated fashion while avoiding contamination by improper storage.
Manufacturer:  Eppendorf™ 4880000000
 View more versions of this product
Catalog No. 13-690-210
Description
Description
Store Eppendorf pipettes and/or liquid handling instruments in a comfortable and safe way with this pipette carousel. Eppendorf™ Electronic Pipet Charging Stands charge electronic pipettes in an uncomplicated fashion while avoiding contamination by improper storage.
  • When used with an additional charger shell, the charger stand is also perfect for the Repeater stream™ and Repeater Xstream™/Repeater™ E3 and E3x
  • Quadruple charging rack
  • For Multipette Stream/Xstream pipets
Order Info
Does not include power supply.
Specifications
Specifications
Charging Stand, 1 Pipet
Xplorer™ electronic pipette
Documents