Eppendorf™ Electronic Pipet Charging stands
Store 4 Eppendorf pipettes and/or liquid handling instruments in a comfortable and save way with this pipette carousel. Eppendorf™ Electronic Pipet Charging Stands charge electronic pipettes in an uncomplicated fashion while avoiding contamination by improper storage.
$453.07 - $1,413.83
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
13-690-210 Eppendorf™
4880000000
Xplorer™ electronic pipette Each for $481.31
13-690-211 Eppendorf™
4880000026
Xplorer™ electronic pipette Each for $1,413.83
36-102-5705 Eppendorf™
4880000018
Repeater™ stream and Repeater™ Xstream pipette Each for $453.07
Specifications
Description
  • When used with an additional charger shell, the charger stand is also perfect for the Repeater stream™ and Repeater Xstream™/Repeater™ E3 and E3x
  • Quadruple charging rack
  • For Multipette Stream/Xstream pipets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
0.0