Eppendorf™ Combitips Advanced™ Pipetter Tips - Standard, Sterile
High-precision and quick dispensing independent of liquid density and viscosity
Manufacturer:  Eppendorf™ 0030089510
 View more versions of this product
Catalog No. 13-683-709
Description
Description
Combitips advanced are the ideal components for all Multipettes™. Working with radioactive, aggressive or toxic materials also becomes safer due to hermetically sealed piston preventing aerosol contamination. The new individual color coding facilitates quick identification of the desired tip.
  • Slip agent free
  • Color-coded tip size recognition
  • Funnel geometry from cylinder to rim; prevents damage to gloves and ensures comfortable handling
  • Function according to the positive displacement principle
  • Elongated design for complete emptying of all common tubes
  • Optimized graduations for visibility of liquid volume
  • Automatic Combitip recognition and volume calculation
  • 3D design on the upper rim for easy recognition
  • Maximum range offered by nine volume sizes (0.1mL to 50mL)
  • Dust free storage
Specifications
Specifications
Eppendorf™ Multipette™
0.1mL
Yes
Extended Length, Positive Displacement
Sterile
Eppendorf™ Multipette™
Documents