Eppendorf™ Biopur™ Racks and Singles
For the most stringent laboratory requirements
Manufacturer:  Eppendorf™ 022491164
 View more versions of this product
Catalog No. 05-403-118
Description
Description
  • Sterile; RNase-, DNA-, ATP- and pyrogen-free
  • Drop contour design provide better tip sealing
  • Finely tapered and extended; ideal for small-volume pipetting
  • Fine graduations make it easy to check volume
  • Soft, flexible tip cone provides better seal to pipetter
  • Manufactured under strict quality standards in ISO-9001-certified production process
  • Available in racks or individually wrapped
Specifications
Specifications
Rack
1 to 10mL
Turquoise
6.49 in.
ISO 9001
Universal Fit
Sterile
PP (Polypropylene)
16.5cm
Documents