missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ 5810R Centrifuge and Rotor Packages

The workhorse of the lab with user-friendly operation and refrigeration for maximum versatility

Manufacturer:  Eppendorf™ 022628169

 View more versions of this product

Catalog No. 05-413-401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Includes: Centrifuges, rotors, and clinical and cell-culture packages

Description

Description

For medium to high through-put applications including high capacity for cell culture and clinical applications, high-speed for separating cell lysates and a microcentrifuge for nucleic acid purifications.

  • Speed, rcf, and radius correction values can be entered by the user
  • Values can be changed during centrifugation
  • Speed can be set from 220-14,000rpm (20,817xG) to maximum speed in increments of 10 rpm
  • Selectable program memory for up to 35 individual user programs
  • Adjustable speed short spin button for quick runs
  • “at set rpm” for even more reproducable results
  • 10 acceleration and 10 braking ramps for sensitive sample material
  • Low-profile 11.2 in/28 cm access height for easy sample loading and unloading
  • Automatic rotor recognition senses rotor type to set maximum allowable speedensures safety—fast, simple programming of time, temperature (refrigerated models), speed/ g-force and radius value

Refrigeration Features

  • Temperature range from -9 to +40°C
  • Standby cooling maintains temperature when not in use
  • FastTemp function for fast pre-cooling in as little as 15 minutes and maximum temperature accuracy of the rotor
  • ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life
  • Built-in condensation drain to eliminate water accumulation and prevent corrosion

Order Info

Max. power requirement: 1650w. Power supply: 230V 50/60Hz. Dimensions (L × W × H): 24.0 × 27.6 × 13.8 in. (61 × 70 × 35cm). Weight: 218 lb. (99 kg).

Specifications

Specifications

10 acceleration and 10 braking ramps for sensitive sample material
Selectable program memory for up to 35 individual user programs
Series 5108R Package
14,000rpm
13.8 in.
24 in.
27.6 in.
1650w
120V 50/60Hz
5810R
218 lb. (99kg)
-9° to +40°C
20,817 x G
Yes
35cm
61cm
70cm
4 × 250mL rotor, 15mL and 50mL adapters (1pk.)
Yes
Documents