missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ 5810R Centrifuge and Rotor Packages

The workhorse of the lab with user-friendly operation and refrigeration for maximum versatility

Manufacturer:  Eppendorf™ 022628168

 View more versions of this product

Catalog No. 05-413-315

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Includes: Centrifuges, rotors, and clinical and cell-culture packages

Description

Description

For medium to high through-put applications including high capacity for cell culture and clinical applications, high-speed for separating cell lysates and a microcentrifuge for nucleic acid purifications.

  • Speed, rcf, and radius correction values can be entered by the user
  • Values can be changed during centrifugation
  • Speed can be set from 220-14,000rpm (20,817xG) to maximum speed in increments of 10 rpm
  • Selectable program memory for up to 35 individual user programs
  • Adjustable speed short spin button for quick runs
  • “at set rpm” for even more reproducable results
  • 10 acceleration and 10 braking ramps for sensitive sample material
  • Low-profile 11.2 in/28 cm access height for easy sample loading and unloading
  • Automatic rotor recognition senses rotor type to set maximum allowable speedensures safety—fast, simple programming of time, temperature (refrigerated models), speed/ g-force and radius value

Refrigeration Features

  • Temperature range from -9 to +40°C
  • Standby cooling maintains temperature when not in use
  • FastTemp function for fast pre-cooling in as little as 15 minutes and maximum temperature accuracy of the rotor
  • ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life
  • Built-in condensation drain to eliminate water accumulation and prevent corrosion

Order Info

Max. power requirement: 1650w. Power supply: 230V 50/60Hz. Dimensions (L × W × H): 24.0 × 27.6 × 13.8 in. (61 × 70 × 35cm). Weight: 218 lb. (99 kg).

Specifications

Specifications

10 acceleration and 10 braking ramps for sensitive sample material
Selectable program memory for up to 35 individual user programs
1L
20,817 x G
Yes
35cm
61cm
70cm
4 × 250mL Swinging Bucket Rotor and Adapter Bundle
Yes
218 lb. (99kg)
-9° to +40°C
Series 5108R Package
14,000rpm
13.8 in.
24 in.
27.6 in.
1650w
120V 50/60Hz
5810R
Documents