Eppendorf™ 5702 Series Fixed-Angle Rotors
For use with 5702 centrifuge
Supplier: Eppendorf™ 022639404
Includes: 05-400-304 includes 30 steel adapter sleeves, 20 cushions for conical tubes and 30 cushions for round-bottom tubes. 05-400-305 includes 10 steel adapter sleeves, 10 cushions for conical tubes and 10 cushions for 15mL round-bottom tubes.
Description
- 30 or 10 × 15mL capacity
- Max. speed/force: 4400rpm/2750 x g
Specifications
Fixed Angle Rotor | |
2,750 x g | |
5702 Centrifuge |
30 x 15 mL | |
4,400 rpm | |
30 Adapters and 30 Rubber mats, 30 Steel adapter sleeves, 20 cushions for conical tubes and 30 cushions for round bottom tubes |