missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ 5702 Series Centrifuge

Low speed, general purpose units designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs

Manufacturer:  Eppendorf™ 022628023

 View more versions of this product

Catalog No. 05-413-320

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Choose either the 5702 model - a base, non-heated centrifuge for standard applications, the 5702 R that is a refrigerated model for heat-sensitive samples, or the heated and refrigerated 5702 RH model for centrifugation under biological conditions to improve cell viability for more accurate results in downstream application like cell cultivation.
Features

  • Max. speed: 3,000 x g (4,400 rpm)
  • Very compact footprint to fit on every lab bench
  • Very quiet operation to benefit your environment
  • Low access height for simple loading and unloading of samples
  • SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
  • “At set RPM” function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
  • Key lock to prevent unintentional adjustments
  • Stainless steel rotor chamber is rust-free and easy to clean
  • Electronic imbalance detection for maximum safety
  • Optimizes air flow minimizes sample warming during run (5702)
  • FastTemp function for fast precooling (5702 R/RH)
  • Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
  • ECO shut–off engages after 8 hours of nonuse to reduce energy consumption and extend compressor life (5702 R/RH)
  • Stainless steel rotor chamber is rust-free and easy to clean
  • Imbalance detection for maximum safety
  • Key lock to prevent accidental parameter changes
  • Aerosol–tight caps for safe centrifugation of hazardous samples optionally available

Additional Features for 5702 R and 5702 RH
  • Temperature settings from -9 to 40°C
  • FastTemp™ function for fast precooling
  • Standby cooling keeps the 5702 R at set temperature when lid is closed
  • ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life
  • Two program keys to store routine procedures
  • Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)

Packages
All packages include one centrifuge and a rotor. Other parts included may be a bucket, adapters, and rotary knobs. See ordering table for specifics of each package.
Specifications

Specifications

25 s/25 s
0 – 99 min, with continuous run function
4 x 100mL
3,000 x G
60cm (23.6 in.)
SOFT brake function
Centrifuge 5702 RH, refrigerated/heated, with Rotor A-4-38 incl. adapters for 13/16 mm blood collection tubes, rotary knobs, 120 V/50 – 60 Hz (US)
5702RH
79.4 lb. (36kg)
-9° to +40°C
Refrigerated/Heated
4,400rpm
< 45 dB(A) with Rotor A-4-38
380w
120V 50/60Hz
Documents