missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ 5702 Series Centrifuges Packages

Package provides centrifuge plus rotor—some models also include four adapters for clinical and cell culture applications

$5,420.76 - $5,420.76

Specifications

Certifications/Compliance CE, UL, cUL, IVD requirements
Controller Type Large Dial
Dimensions (L x W x H) Exterior 15.7 x 12.6 x 9.4 in. (40 x 32 x 24cm)
Hertz 60Hz
Max. RCF 3000 x G
View More Specs

Includes: Centrifuges, rotors, and adapters (selected models)


Products ${productFamilyLength}
Catalog Number Mfr. No. Electrical Requirements Height (Metric) Exterior Length (Metric) Exterior Product Type Price Quantity  
Catalog Number Mfr. No. Electrical Requirements Height (Metric) Exterior Length (Metric) Exterior Product Type Price Quantity  
05-413-300 Eppendorf™
022628001
120V 60Hz 24cm 40cm Model 5702 Each for $5,420.76
Specifications

Description

All 5702 models are available in standard, refrigerated, and refreigerated/heated versions, and can be used in a range of settings, from clinical to cell culture labs.

Common Features

  • Very compact design
  • Low access height
  • Automatic motorized locking of the lid
  • Slow startup and braking through the SOFT function
  • Stainless-steel, rustproof rotor chamber
  • Optimized airflow directed backwards

Model 5702 (no temperature control)

  • Switch between rpm and rcf
  • Speed set from 100 rpm to maximum of 4400rpm (3000xG), in increments of 100rpm
  • Separate Short Spin button
  • Timer can be set for up to 99 minutes
  • Continuous mode can be selected
  • Parameter locking
  • Super quiet
  • “At Set” function starts timer when set speed is reached for reproducible results
  • Glass window for quick calibration check
  • Six rotor options
  • SOFT brake function for delicate samples
  • Optimized braking ramp for isolation of mononuclear blood cells using Ficoll density gradient centrifugation
Model 5702 R (refrigerated) - Additional Features
  • Adjustable from -9° to +40°C
  • Standby refrigeration
  • “At temp” function - quickly reach set temperature
  • ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life
  • Built-in condensation drain to eliminate water accumulation and prevent corrosion
  • Use for safe holding at 4°C
Model 5702 RH (refrigerated & heated)
  • Adjustable from -9° to +43°C
  • Immediately ready for use via standby heating/refrigeration
  • Temperature holds steady at 4°±1°C and 37°C±1°C
  • Active heating
  • Precise temperature control, software controlled depending on rotor
  • Two programming keys to store routine procedures
  • Ideal for conjugation of sperm in IVF labs and spin-down of temperative-sensitive active humam cells
Included 4 × 100mL Swing-Bucket Rotors
  • Aluminum, round buckets with aerosol-tight caps available
  • Maximum rotational speed: 4400rpm (maximum rcf: 3000xG)
  • Square buckets available for use with glass, round-bottom tubes

Included Adapters (selected models)
  • 4 × 13mm O.D. adapters and 4 × 16mm O.D. adapters
  • 4 ×15 and 4 × 50mL Conical adapters

Order Info

Max. power requirements, Model 5702: 200w; Models 5702 R, RH: 380w. Power supply, all models: 230V 50Hz. Dimensions (L × W × H), Model 5702: 15.7 × 12.6 × 9.4 in. (40 × 32 × 24cm); Models 5702 R, RH: 22.8 × 15.0 × 10.2 in. (58 × 38 × 26cm). Weight, Model 5702: 44.0 lb. (20kg); Models 5702 R, RH: 79.4 lb. (36kg).

Compliance

Conform to CE, UL, cUL, IVD requirements

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

CE, UL, cUL, IVD requirements
15.7 x 12.6 x 9.4 in. (40 x 32 x 24cm)
3000 x G
Super Quiet
Up to 99 min.
Large Dial
60Hz
400rpm
No
No