Please login to your online account to display your discounted pricing

Eppendorf™ 5702 Series Centrifuge


Low speed, general purpose units designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs

Manufacturer: eppendorf™  022628012

 View more versions of this product

Catalog No. 05-413-301

  • / Each
 

Description & Specifications

Specifications

Profile (Acceleration/Braking) 25 s/25 s
Temperature Range (Metric) -9° to +40°C
Max. Speed 4400rpm
Noise Level < 45 dB(A) with Rotor A-4-38
Capacity (Metric) Max. Load 4 x 100mL
Height (Metric) Open Cover 60cm (23.6 in.)
Safety Features SOFT brake function
Timer 0 – 99 min, with continuous run function
Includes Centrifuge 5702 R, refrigerated, refrigerated, with Rotor A-4-38 incl. adapters for 13/16 mm blood collection tubes, rotary knobs, 120 V/50 – 60 Hz (US)
Electrical Requirements 120V 50-60Hz
Dimensions (L x W x H) Exterior 58 x 38 x 26cm (22.8 x 15 x 10.2 in.)
Model 5702R
Product Type Refrigerated
Weight (English) 79.4 lb.
Max. RCF 3000 x G
Wattage 380w
Weight (Metric) 36kg

Choose either the 5702 model - a base, non-heated centrifuge for standard applications, the 5702 R that is a refrigerated model for heat-sensitive samples, or the heated and refrigerated 5702 RH model for centrifugation under biological conditions to improve cell viability for more accurate results in downstream application like cell cultivation.
Features

  • Max. speed: 3,000 x g (4,400 rpm)
  • Very compact footprint to fit on every lab bench
  • Very quiet operation to benefit your environment
  • Low access height for simple loading and unloading of samples
  • SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
  • “At set RPM” function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
  • Key lock to prevent unintentional adjustments
  • Stainless steel rotor chamber is rust-free and easy to clean
  • Electronic imbalance detection for maximum safety
  • Optimizes air flow minimizes sample warming during run (5702)
  • FastTemp function for fast precooling (5702 R/RH)
  • Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
  • ECO shut–off engages after 8 hours of nonuse to reduce energy consumption and extend compressor life (5702 R/RH)
  • Stainless steel rotor chamber is rust-free and easy to clean
  • Imbalance detection for maximum safety
  • Key lock to prevent accidental parameter changes
  • Aerosol–tight caps for safe centrifugation of hazardous samples optionally available

Additional Features for 5702 R and 5702 RH
  • Temperature settings from -9 to 40°C
  • FastTemp™ function for fast precooling
  • Standby cooling keeps the 5702 R at set temperature when lid is closed
  • ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life
  • Two program keys to store routine procedures
  • Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)

Packages
All packages include one centrifuge and a rotor. Other parts included may be a bucket, adapters, and rotary knobs. See ordering table for specifics of each package.