missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ 5702 Series Centrifuge
Low speed, general purpose units designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs
Supplier: Eppendorf™ 022628124DUP
Description
Choose either the 5702 model - a base, non-heated centrifuge for standard applications, the 5702 R that is a refrigerated model for heat-sensitive samples, or the heated and refrigerated 5702 RH model for centrifugation under biological conditions to improve cell viability for more accurate results in downstream application like cell cultivation.Features
- Max. speed: 3,000 x g (4,400 rpm)
- Very compact footprint to fit on every lab bench
- Very quiet operation to benefit your environment
- Low access height for simple loading and unloading of samples
- SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
- “At set RPM” function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
- Key lock to prevent unintentional adjustments
- Stainless steel rotor chamber is rust-free and easy to clean
- Electronic imbalance detection for maximum safety
- Optimizes air flow minimizes sample warming during run (5702)
- FastTemp function for fast precooling (5702 R/RH)
- Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
- ECO shut–off engages after 8 hours of nonuse to reduce energy consumption and extend compressor life (5702 R/RH)
- Stainless steel rotor chamber is rust-free and easy to clean
- Imbalance detection for maximum safety
- Key lock to prevent accidental parameter changes
- Aerosol–tight caps for safe centrifugation of hazardous samples optionally available
Additional Features for 5702 R and 5702 RH
- Temperature settings from -9 to 40°C
- FastTemp™ function for fast precooling
- Standby cooling keeps the 5702 R at set temperature when lid is closed
- ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life
- Two program keys to store routine procedures
- Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
Packages
All packages include one centrifuge and a rotor. Other parts included may be a bucket, adapters, and rotary knobs. See ordering table for specifics of each package.
Specifications
4 x 100 mL | |
4,400 rpm | |
<45 dB(A) with Rotor A-4-38 | |
0 to 99 min., with continuous run function | |
-9°C to +40°C | |
10.2 x 15 x 22.8 in. (26 x 38 x 58 cm) | |
79.4 lb. (36 kg) | |
380 w |
3,000 x g | |
Centrifuge 5702 RH; Refrigerated/Heating; with 4 x 100 mL Swing Bucket Rotor, 4 x 15 mL Adapters, 4 x 50 mL Adapters (Cell Culture) | |
25/25 sec. | |
SOFT brake function | |
Model 5702 RH | |
60 cm | |
230 V 60 Hz |