missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ 30-Place Fixed-Angle Rotors

For Models 5417C/5417R Microcentrifuges

Manufacturer:  Eppendorf™ 022636006

 View more versions of this product

Catalog No. 05-406-13



Includes: Cover

Description

Description

Ideal for containing hazardous materials such as HIV, TB and radioactive samples.

  • Accept up to 30 × 1.5 to 2.0mL tubes directly or 0.5mL, 0.25/0.4mL, Microtainer™ and 0.2mL PCR tubes with accessory adapters
  • Maximum speed/RCF: 14,000rpm/20,800 x g
  • Available with quick-release lid or aerosol-tight cover

Specifications

Specifications

Fixed Angle Rotor
20,800 x g
Eppendorf™ 5417C/5417R Microcentrifuge
Yes
30 x 1.5/2mL
14,000rpm
Fixed
Aerosol-tight lid
Documents
Accessories