Please login to your online account to display your discounted pricing

Eppendorf™ 0.2mL PCR Tube Strips

Hinged caps open to a 90° angle—lids will not cover adjacent tubes in thermal cycler block

$301.00 - $301.00

Specifications

Capacity (Metric) 0.2mL
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

E0030124286

 
eppendorf™
951010022
Case of 120 Each for $301.00
Description & Specifications

Specifications

Capacity (Metric) 0.2mL

  • Clear polypropylene with etched writing surface and snap cap closures
  • Thin walls ensure uniform, rapid heat transfer in thermal cycler blocks
  • Integral contamination shield prevents fingertip from touching inner lid surface when opening
  • Certified free of human DNA, DNase, RNase, and PCR inhibitors
  • Autoclavable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.