Please login to your online account to display your discounted pricing

Alfa Aesar™ Epoxide functional resin, ca 2 mmol/g

Chemical Identifiers

Chemical Name or Material Epoxide functional resin
MDL Number MFCD03456957
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1959703

  SDS
alfa aesar™
L1959703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1959706

  SDS
alfa aesar™
L1959706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1959714

  SDS
alfa aesar™
L1959714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Epoxide functional resin
MDL Number MFCD03456957
Concentration ≈2mmol/g
Quantity 1g