Please login to your online account to display your discounted pricing

Eplerenone, Tocris

Selective mineralocorticoid receptor antagonist

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material Eplerenone
Assay Percent Range >99%
Molecular Formula C24H30O6
Formula Weight 414.5
Synonym (7α,11α,17α)-9,11-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic acid γ-lactone methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

239710

 
r & d systems
RDS239710
10mg Each for $195.49

239750

 
r & d systems
RDS239750
50mg Each for $816.90
Description & Specifications

Specifications

Formula Weight 414.5
Quantity 10mg
Chemical Name or Material Eplerenone
Molecular Formula C24H30O6
CAS 107724-20-9
Synonym (7α,11α,17α)-9,11-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic acid γ-lactone methyl ester
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature