Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-3, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and radioimaging experiments.

Manufacturer: thermo scientific™ lab vision™  MS181P1ABX

 View more versions of this product

Catalog No. MS181P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen Epithelial Specific Antigen/Ep-CAM Ab-3
Immunogen MCF-7 human breast cancer cells
Clone 323/A3
Host Species Mouse
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Quantity 100µL
Format Concentrated
Isotype IgG1
Type Primary
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Research Discipline Cancer and Tumor Biology

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and radioimaging experiments with Thermo Scientific™ Epithelial Specific Antigen/Ep-CAM Ab-3, Mouse Monoclonal Antibody.

EGP40 is a 40 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.

Host Species: Mouse

Clone: 323/A3

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: MCF-7 human breast cancer cells

Molecular Weight: 40-43 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
  • Radioimaging

Note:

USA: RUO; Int'l: RUO