Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-2, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Epithelial Specific Antigen/Ep-CAM Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS675P1ABX

 View more versions of this product

Catalog No. MS675P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen Epithelial Specific Antigen/Ep-CAM Ab-2
Host Species Mouse
Isotype IgG2a kappa
Quantity 100µL
Clone AUA1
Immunogen Colon carcinoma LoVo cell line
Format Concentrated
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cancer and Tumor Biology
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Classification Monoclonal
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Epithelial Specific Antigen/Ep-CAM Ab-2, Mouse Monoclonal Antibody.

EGP40 is a 40 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.

Host Species: Mouse

Clone: AUA1

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Colon carcinoma LoVo cell line

Molecular Weight: 40-43 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO