Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-5, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

$262.29 - $1,295.12

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-5
Clone H11
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS316P

 
thermo scientific™ lab vision™
MS316P
Purified Ab with BSA and Azide 1mL Each for $1,218.04

MS316P0

 
thermo scientific™ lab vision™
MS316P0
Purified Ab with BSA and Azide 100µL Each for $262.29

MS316P1

 
thermo scientific™ lab vision™
MS316P1
Purified Ab with BSA and Azide 500µL Each for $725.04

MS316P1ABX

 
thermo scientific™ lab vision™
MS316P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $800.76

MS316PABX

 
thermo scientific™ lab vision™
MS316PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,295.12
Description & Specifications

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-5
Clone H11
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen HC2 20 d2 cells
Isotype IgG1 kappa

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-5, Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: H11

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: HC2 20 d2 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells, Placenta or Squamous cell CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO