Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-15, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments.

$271.55 - $1,367.59

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-15
Clone H9B4
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS665P

 
thermo scientific™ lab vision™
MS665P
200μg/mL 1mL Each for $1,270.55

MS665P0

 
thermo scientific™ lab vision™
MS665P0
200μg/mL 100µL Each for $271.55

MS665P1

 
thermo scientific™ lab vision™
MS665P1
200μg/mL 500µL Each for $769.67

MS665P1ABX

 
thermo scientific™ lab vision™
MS665P1ABX
1mg/mL 100µL Each for $849.53

MS665PABX

 
thermo scientific™ lab vision™
MS665PABX
1mg/mL 200µL Each for $1,367.59
Description & Specifications

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-15
Clone H9B4
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen Purified EGFR from A431 cells
Isotype IgG1

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-15, Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: H9B4

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Cytoplasmic domain

Immunogen: Purified EGFR from A431 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO