Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-15, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS665P1ABX

 View more versions of this product

Catalog No. MS665P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-15
Applications Immunoprecipitation
Classification Monoclonal
Clone H9B4
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen Purified EGFR from A431 cells
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-15, Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: H9B4

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Cytoplasmic domain

Immunogen: Purified EGFR from A431 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO