Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-13, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunoprecipitation experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-13, Mouse Monoclonal Antibody.

$271.20 - $1,583.24

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-13
Clone 528 + 199.12
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS609B

 
thermo scientific™ lab vision™
MS609B
200μg/mL Biotin 1mL Each for $1,583.24

MS609B0

 
thermo scientific™ lab vision™
MS609B0
200μg/mL Biotin 100µL Each for $366.94

MS609B1

 
thermo scientific™ lab vision™
MS609B1
200μg/mL Biotin 500µL Each for $998.35

MS609P

 
thermo scientific™ lab vision™
MS609P
200μg/mL Unlabeled 1mL Each for $1,280.27

MS609P0

 
thermo scientific™ lab vision™
MS609P0
200μg/mL Unlabeled 100µL Each for $271.20

MS609P1

 
thermo scientific™ lab vision™
MS609P1
200μg/mL Unlabeled 500µL Each for $763.94

MS609P1ABX

 
thermo scientific™ lab vision™
MS609P1ABX
1mg/mL Unlabeled 100µL Each for $843.55

MS609PABX

 
thermo scientific™ lab vision™
MS609PABX
1mg/mL Unlabeled 200µL Each for $1,367.98
Description & Specifications

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-13
Clone 528 + 199.12
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombinant human EGFR protein (clone 199.12)
Isotype IgG2a

Obtain accurate, reproducible results in immunoprecipitation experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-13, Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 528 + 199.12

Isotype: IgG2a + IgG2a

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombinant human EGFR protein (clone 199.12)

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO