Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-12 (Cocktail R19/48), Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments.

$312.31 - $1,415.39

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-12 (Cocktail R19/48)
Clone 111.6+199.12+ F4+11E8
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS400P

 
thermo scientific™ lab vision™
MS400P
200μg/mL 1mL Each for $1,415.39

MS400P0

 
thermo scientific™ lab vision™
MS400P0
200μg/mL 100μL Each for $312.31

MS400P1

 
thermo scientific™ lab vision™
MS400P1
200μg/mL 500μL Each for $983.55

MS400P1ABX

 
thermo scientific™ lab vision™
MS400P1ABX
1mg/mL 100μL Each for $992.47
Description & Specifications

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-12 (Cocktail R19/48)
Clone 111.6+199.12+ F4+11E8
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen Extracellular and cytoplasmic domains of recombinant human EGFR protein
Isotype IgG1 IgG2a

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-12 (Cocktail R19/48), Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 111.6 + 199.12 + F4 + 11E8

Isotype: IgG1 + IgG2a

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Extracellular and cytoplasmic domain

Immunogen: Extracellular and cytoplasmic domains of recombinant human EGFR protein

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native only)

Note:

USA: RUO; Int'l: RUO