Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-10, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

$257.98 - $1,505.41

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-10
Clone 111.6
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS378B

 
thermo scientific™ lab vision™
MS378B
Biotinylated Ab with BSA and Azide 1mL Each for $1,505.41

MS378B0

 
thermo scientific™ lab vision™
MS378B0
Biotinylated Ab with BSA and Azide 100µL Each for $345.35

MS378B1

 
thermo scientific™ lab vision™
MS378B1
Biotinylated Ab with BSA and Azide 500µL Each for $949.33

MS378P

 
thermo scientific™ lab vision™
MS378P
Purified Ab with BSA and Azide 1mL Each for $1,199.61

MS378P0

 
thermo scientific™ lab vision™
MS378P0
Purified Ab with BSA and Azide 100µL Each for $257.98

MS378P1

 
thermo scientific™ lab vision™
MS378P1
Purified Ab with BSA and Azide 500µL Each for $722.35

MS378P1ABX

 
thermo scientific™ lab vision™
MS378P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $795.88

MS378PABX

 
thermo scientific™ lab vision™
MS378PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,298.18

MS378R7

 
thermo scientific™ lab vision™
MS378R7
Ready-to-use for IHC (UVLP) 7mL Each for $679.08
Description & Specifications

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-10
Clone 111.6
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Immunogen Extracellular domain of human recombinant EGFR protein
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-10, Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 111.6

Isotype: IgG1

Species Reactivity: Human. Not mouse and rat.

Epitope: Extracellular domain

Immunogen: Extracellular domain of human recombinant EGFR protein

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells; Skin, Placenta or Squamous cell CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native only)
  • Effect on EGF Binding (Inhibitory)

Note:

USA: RUO; Int'l: RUO