Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-1, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in affinity purification, immunoprecipitation and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS268P1ABX

 View more versions of this product

Catalog No. MS268P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Cross Reactivity Canine
Cross Reactivity Human
Clone 528
Host Species Mouse
Antigen Epidermal Growth Factor Receptor /EGFR Ab-1
Isotype IgG2a
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunoprecipitation
Quantity 100µL
Immunogen Purified EGFR from A431 cells
Classification Monoclonal
Research Discipline EGFR Family
Concentration 1mg/mL
Conjugate Unlabeled
Type Primary

Obtain accurate, reproducible results in affinity purification, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-1, Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 528

Isotype: IgG2a

Species Reactivity: Human and Dog. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: Purified EGFR from A431 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Affinity Purification
  • Blocks EGF/TGFα-induced Activation of EGFR
  • Inhibits Binding of EGFR Ab-2 (clone 225) to EGFR
  • Inhibits Proliferation of A431 Cells
  • Arrests Tumor Growth in vivo
  • Immunoprecipitation (Native only)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO