Please login to your online account to display your discounted pricing

Alfa Aesar™ (-)-Epicatechin

An antioxidant and natural product from green tea

Chemical Identifiers

Chemical Name or Material (-)-Epicatechin
CAS 490-46-0
Formula Weight 290.27
MDL Number MFCD00075648
Beilstein 92760
Synonym (-)-cis-3,3',4',5,7-Pentahydroxyflavane; Epicatechol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ61218FPL

  SDS
alfa aesar™
J61218FPL
20mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 240°C (dec.)
Formula Weight 290.27
Chemical Name or Material (-)-Epicatechin
CAS 490-46-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00075648
Beilstein 92760
Physical Form Crystalline Powder
Storage Note 1 2° to 8°C
RTECS Number KB3745000
Quantity 20mg
Color White to pale yellow
Health Hazard 1 Warning
Synonym (-)-cis-3,3',4',5,7-Pentahydroxyflavane; Epicatechol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Solubility Information Soluble in water or alcohol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C