EPHB4 Recombinant Human Protein DFS Item
EPHB4 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3251
 View more versions of this product
Catalog No. PV3251
Description
Description
EPHB4 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation.
Specifications
Specifications
NP_004435
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
50.5kDa
95% by SDS-PAGE
–80°C
1 Each
EPHB4, TYRO11
EPHB4 was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MgCl2, 2.5mM DTT, 0.01% Triton™ X-100, 10% glycerol, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
Ephrin and EPH receptor
EPHB4 Recombinant Human Protein
Human
0.35mg/mL
2050
EPHB4 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Receptor tyrosine kinases
EPHB4 Recombinant Human Protein
Kinase assay
Baculovirus
Amino acids 561-987
400nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
His Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.