Invitrogen™ EPHB3 Recombinant Human Protein DFS Item
EPHB3 is a receptor tyrosine kinase that belongs to the ephrin receptor family.
Manufacturer:  Invitrogen™ LSPV4102
 View more versions of this product
Catalog No. PV4102
Description
Description
Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation.
Specifications
Specifications
NP_004434
Frozen
37.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
EPH Receptor Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
EPHB3, HEK2, TYRO6
Baculovirus
Human
GST Tag
Documents