Invitrogen™ EPHB1 Recombinant Human Protein DFS Item
EPHB1 is a receptor tyrosine kinase that belongs to the ephrin receptor family.
Manufacturer:  Invitrogen™ LSPV4100
 View more versions of this product
Catalog No. PV4100
Description
Description
Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation.
Specifications
Specifications
NP_004432
Frozen
60.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
EPH Receptor Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
EPHB1, HEK6
Baculovirus
Human
GST Tag
Documents