missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
EPHB1 Recombinant Human Protein
EPHB1 Recombinant Human Protein
Supplier: Thermo Fisher Scientific PV3786
Description
EPHB1 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation.Specifications
NP_004432 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
60.3kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
EPHB1, HEK6 | |
EPHB1 was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 612-887 | |
645nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
GST Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
0.49mg/mL | |
2047 | |
EPHB1 Recombinant Human Protein | |
–80°C | |
1 Each | |
EPHB1 Recombinant Human Protein | |
Ephrin and EPH receptor | |
EPHB1 Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.