missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ EPHA8 Recombinant Human Protein DFS Item

EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family.

$2,040.00 - $8,670.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR7298B
View Documents
Invitrogen™
LSPR7298B
100μg Each for $2,040.00
 
PV4099
View Documents
Invitrogen™
LSPV4099
1mg Each for $8,670.00
 
Description

Description

EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system.

Specifications

Specifications

Frozen
Human
GST Tag
Yes