Invitrogen™ EPHA8 Recombinant Human Protein DFS Item
EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family.
Manufacturer:  Invitrogen™ LSPV4099
 View more versions of this product
Catalog No. PV4099
Description
Description
EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system.
Specifications
Specifications
NP_065387
Kinase Assay
Frozen
76.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
EPH Receptor Kinase Family
Yes
Baculovirus
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
EPHA8, HEK6
Human
GST Tag