EPHA8 Recombinant Human Protein DFS Item
EPHA8 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3844
 View more versions of this product
Catalog No. PV3844
Description
Description
EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system.
Specifications
Specifications
NP_065387
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
76.7kDa
75% by SDS-PAGE
–80°C
1 Each
EPHA8, HEK6
EPHA8 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
Ephrin and EPH receptor
EPHA8 Recombinant Human Protein
Human
0.38mg/mL
2046
EPHA8 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Receptor tyrosine kinases
EPHA8 Recombinant Human Protein
Kinase assay
Baculovirus
Amino acids 565-1005
58nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL
GST Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.