Invitrogen™ EPHA7 Recombinant Human Protein DFS Item
EPHA7 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family.
Manufacturer:  Invitrogen™ LSPV4098
 View more versions of this product
Catalog No. PV4098
Description
Description
EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA7 is involved in embryonic development.
Specifications
Specifications
NP_004431.1
Frozen
76.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
EPH Receptor Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
EPHA7, HEK11, EHK3
Baculovirus
Human
GST Tag
Documents