Learn More
Invitrogen™ EPHA6 Recombinant Human Protein
EPHA6 is a member of the ephrin receptor subfamily of protein-tyrosine kinases. Recombinant human EPHA6 (561-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Manufacturer: Invitrogen™ LSPV6338
Description
EPHA6 has been implicated in axon guidance, neuron-target interactions, regional compartmentalization, and synaptic functions in nervous systems. EPHA6 is highly expressed in the brain and testis. Reduction in EPHA6 has been detected in Hypospadias, a common defect affecting the growth and closure of the external genitalia. Genetic inhibition of EPHA6 in mice produces behavioral deficits specifically in tests of learning and memory. Mice deficient in EPHA6 show reduced memory of the consequences of the training context.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma | |
77.0kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
Kinase Assay | |
Human | |
GST Tag |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol | |
100μg | |
EPA6, FLJ35246, PRO57066, DKFZp434C1418 | |
aa 561-end | |
Yes |