Invitrogen™ EPHA2 Recombinant Human Protein DFS Item
EPHA2 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family.
Manufacturer:  Invitrogen™ LSPV4094
 View more versions of this product
Catalog No. PV4094
Description
Description
EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA2 is overexpressed in many types of human cancers.
Specifications
Specifications
NP_004422.2
Frozen
76.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
EPH Receptor Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
EPHA2, ECK
Baculovirus
Human
GST Tag
Documents