missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
EPB41L2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002037B01P
Description
Sequence: MTTEVGSVSEVKKDSSQLGTDATKEKPKEVAENQQNQSSDPEEEKGSQPPPAAESQSSLRRQKREKETSESRGISRFIPPWLKKQKSYTLVVAKDGGDKKEPTQAVVEEQVLDKEEPLPEEQRQAKGDAEEMAQKKQEIKVEVKEEKPSVSKEEKPSVSKVEMQPTELVSKEREEKVKETQEDKLEGGAAKRETKEVQTNELKAEKASQKVTKKTKTVQCKVTLLDGTEYSCDLEKHAKGQVLFDKVCEHLNLLEKDYFGLLFQESPEQKNWLDPAKEIKRQLRNLPWLFTFNVKFYPPDPSQLTEDITRYFLCLQLRQDIASGRLPCSFVTHALLGSYTLQAELGDYDPEEHGSIDLSEFQFAPTQTKELEEKVAELHKTHRGLSPAQADSQFLENAKRLSMYGVDLHHAKDSEGVDIKLGVCANGLLIYKDRLRINRFAWPKILKISYKRSNFYIKVRPAELEQFESTIGFKLPNHRAAKRLWKVCVEHHTFYRLVSPEQPPKAKFLTLGSKFRYSGRTQAQTRQASTLIDRPAPHFERTSSKRVSRSLDGAPIGVMDQSLMKDFPGAAGEISAYGPGLVSIAVVQDGDGRREVRSPTKAPHLQLIEGKSSHETLNIVEEKSGQRLGKTKESpecifications
EPB41L2 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
BC034718.1 | |
EPB41L2 | |
EPB41L2 (AAH34718.1, 1 a.a. ∼ 633 a.a) full-length human protein. | |
RUO | |
2037 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunofluorescence,Western Blot | |
Unconjugated | |
erythrocyte membrane protein band 4.1-like 2 | |
4.1-G/DKFZp781D1972/DKFZp781H1755 | |
Mouse | |
50 μg | |
Primary | |
Human,Mouse | |
Antibody |